missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Thermo Scientific Splitless FocusLiner (For Use with 70mm Needle)
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 09204501
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Thermo Scientific instruments
Specifications
70 mm Needle | |
1 Ea. | |
5 mm | |
Thermo Scientific Trace and Focus GCs |
GC Inlet LinerSplitless FocusLiner | |
105 mm | |
8 mm |