missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall™ Culture Flask Adapters

For processing Nunc™ T-75 or T-25 EasyFlasks™ in Thermo Scientific Sorvall Legend™ T/RT Centrifuges.

Brand:  Thermo Scientific™ 75008384

Code : NEW

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12613553

  • £720.00 / Pack of 2
Estimated Shipment: 09-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Requires : Rotor 75006445 with round bucket 75006441

Description

Description

Unique angled design ensures tight pelletization, allowing easy removal of supernatant without pellet disruption.

  • T-25 adapter holds flask at 25° angle; T-75 at 75°
  • Sold in pairs

Specifications

Specifications

T-25 Flask
Thermo Scientific™ Sorvall™ Legend™ T/RT rotor with round bucket
25°
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers