Learn More
Thermo Scientific™ Sorvall™ Culture Flask Adapters
For processing Nunc™ T-75 or T-25 EasyFlasks™ in Thermo Scientific Sorvall Legend™ T/RT Centrifuges.
£720.00 - £730.00
Specifications
For Use With (Equipment) | Thermo Scientific™ Sorvall™ Legend™ T/RT rotor with round bucket |
---|
Product Code | Brand | Tube Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Type | Price | Quantity & Availability | |||||
12693523
|
Thermo Scientific™
75008383 |
T-75 Flask |
£730.00
Pack of 2 |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12613553
|
Thermo Scientific™
75008384 |
T-25 Flask |
£720.00
Pack of 2 |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Unique angled design ensures tight pelletization, allowing easy removal of supernatant without pellet disruption.
- T-25 adapter holds flask at 25° angle; T-75 at 75°
- Sold in pairs
Specifications
Thermo Scientific™ Sorvall™ Legend™ T/RT rotor with round bucket |