Learn More
Epredia™ Shandon™ Tissue-Loc™ Tissue Cassettes IV
Compatible with Epredia™ Shandon™ MicroWriter™ and PrintMate™ cassette labelers
Brand:  Epredia™ 41721
Code : A0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12560077
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Disposable
  • Holds tissue specimens during embedding, processing
  • Pore style: Small slots
  • Supplied in box of 250 cassettes with attached lids
Specifications
Specifications
Small Slots
Hinged
Blue