Learn More
Epredia™ Shandon™ Tissue-Loc™ Tissue Cassettes IV
Compatible with Epredia™ Shandon™ MicroWriter™ and PrintMate™ cassette labelers
£59.65 - £64.11
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12560077
|
Epredia™
41721 |
Blue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12570077
|
Epredia™
41722 |
Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12530087
|
Epredia™
41728 |
Lilac |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12550077
|
Epredia™
41720 |
White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Disposable
- Holds tissue specimens during embedding, processing
- Pore style: Small slots
- Supplied in box of 250 cassettes with attached lids
Specifications
Hinged | |
Small Slots |