Learn More
Description
Thermo Scientific Pierce Premium Grade Sulfo-NHS-SS-Biotin is our highest quality formulation of this amine-reactive, cleavable biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount. Sulfo-NHS-SS-biotin is used for cleavable attachment of biotin to protein and cell surface amines.
- High quality - identity and purity confirmed by multiple tests, including quantitative NMR
- Product integrity - enhanced level of testing and characterization compared to standard grade
- Lot retention - ample supply of past lots retained to help ensure future process testing
- Change management - Change Control Notification (CCN) service
- Consistent manufacture - batch-specific manufacturing documentation review
Sulfo-NHS-SS-biotin is a water-soluble, NHS-ester biotinylation reagent with a spacer arm that includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines. This reagent is water-soluble, enabling biotinylation in the absence of organic solvents such as DMSO or DMF. The compound is particularly useful for labeling and purifying cell surface proteins because its sulfonate group prevents it from permeating cell membranes and its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.
Specifications
Specifications
| Product Type | Sulfo NHS-SS-Biotin |
| Quantity | 1 g |
| Content And Storage | Premium Grade Sulfo-NHS ester of biotin with an ethyl-1,3-dithioproprionate spacer. Store with desiccant at -20°C. Sufficient to label a total of 10 g of IgG using typical conditions. |
| Product Line | Pierce |
| Cell Permeability | Cell-Impermeant |
| Chemical Reactivity | Amine |
| Reactive Moiety | Active Ester, Succinimidyl Ester, Sulfo-NHS Ester |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
| Label or Dye | Biotin |
| Spacer | Mid-length, Cleavable |
| Show More |
Frequently Asked Questions (FAQs)
Premium Grade Sulfo NHS-SS-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.