missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Premium Grade Sulfo NHS-SS-Biotin

Product Code. 15332627 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15332627 100 mg 100mg
15342627 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15332627 Supplier Thermo Scientific™ Supplier No. PG82077
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biotin-label antibodies or other proteins and biomolecules in applications where product integrity and risk minimization are paramount with Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-SS-Biotin, a high-quality formulation of this amine-reactive, cleavable biotinylation reagent.

Thermo Scientific Pierce Premium Grade Sulfo-NHS-SS-Biotin is our highest quality formulation of this amine-reactive, cleavable biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount. Sulfo-NHS-SS-biotin is used for cleavable attachment of biotin to protein and cell surface amines.

  • High quality - identity and purity confirmed by multiple tests, including quantitative NMR
  • Product integrity - enhanced level of testing and characterization compared to standard grade
  • Lot retention - ample supply of past lots retained to help ensure future process testing
  • Change management - Change Control Notification (CCN) service
  • Consistent manufacture - batch-specific manufacturing documentation review
Compared to the standard grade product, Premium Grade Sulfo-NHS-SS-Biotin provides more clearly defined quality and product support by including increased analytical testing and product characterization, greater batch-specific information and quality assurance review, extensive lot sample retention, and change control notification.
Sulfo-NHS-SS-biotin is a water-soluble, NHS-ester biotinylation reagent with a spacer arm that includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines. This reagent is water-soluble, enabling biotinylation in the absence of organic solvents such as DMSO or DMF. The compound is particularly useful for labeling and purifying cell surface proteins because its sulfonate group prevents it from permeating cell membranes and its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Sulfo NHS-SS-Biotin
Quantity 100 mg
Content And Storage Premium Grade Sulfo-NHS ester of biotin with an ethyl-1,3-dithioproprionate spacer.
Store with desiccant at -20°C.
Sufficient to label a total of 1 g of IgG using typical conditions.
Product Line Pierce
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Mid-length, Cleavable
Label Type Biotin & Analogs
Show More Show Less
Can you provide the shelf-life for Premium Grade Sulfo NHS-SS-Biotin?

Premium Grade Sulfo NHS-SS-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.