missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Hydrazide-PEG4-Desthiobiotin

Product Code. 15295653 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 x 1 mg
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 15295653

Brand: Thermo Scientific™ 16130

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Label glycoproteins for affinity purification with streptavidin beads

Thermo Scientific EZ-Link Hydrazide-PEG4-Desthiobiotin is a pegylated, carbonyl-reactive labeling reagent whose biotin-like group is elutable from streptavidin, enabling its use for cell surface glycoprotein labeling and purification.

This desthiobiotin variant of biotin is activated to label periodate-oxidized carbohydrates of macromolecules. The hydrazide group conjugates to carbonyls (aldehydes and ketones), such as those created from sialic acid and other sugar components of glycoprotein polysaccharides upon treatment with 1 to 10mM sodium periodate (NaIO4). The desthiobiotin tag binds to streptavidin and other biotin-binding proteins with high specificity yet readily elutes with mild conditions (i.e., by competitive displacement with regular, free biotin). As such, this reagent is a useful alternative to Hydrazide-PEG4-Biotin (Part No. 21360) for avidin-biotin techniques in which nondenaturing elution of the labeled proteins is desired. The hydrophilic, 4-unit polyethylene glycol (PEG) spacer arm enhances solubility and reduces aggregation of labeled proteins stored in solution. The PEG segment also adds length and flexibility to the spacer arm, minimizing steric hindrance involved with binding to avidin molecules. The No-Weigh Format (5 x 1mg resealable vials) eliminates the difficulties associated with weighing small quantities of reagent and maximizes protection of unused reagent from degradation.

Highlights:

  • Desthiobiotin - analog of biotin allows easy elution from streptavidin, an ideal feature for affinity purification applications
  • Glycoprotein labeling - tag glycosylated proteins at sialic acid residues to enable high recovery in pull-down assays with streptavidin beads
  • Cell surface labeling - tag and isolate cell surface glycoproteins; reagent does not permeate membranes of whole cells
  • Aldehyde-reactive - reacts with aldehydes formed by periodate-oxidation of sugar groups
  • Hydrazide-activated - perform reactions at pH 4 to 6 in buffers such as sodium acetate
  • Pegylated - spacer arm contains a hydrophilic, 4-unit, polyethylene glycol (PEG) group
  • Enhances solubility - pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
Desthiobiotin vs. Biotin:
Desthiobiotin is a single-ring, sulfur-free analog of biotin that binds to streptavidin with nearly equal specificity but less affinity than biotin (1/Kd = 10^11 vs. 10^15M, respectively)[1-2]. Consequently, desthiobiotinylated bait proteins and their interacting partners can be eluted readily and specifically from streptavidin affinity resin using mild conditions based on competitive displacement with free biotin. For pull-down assay experiments with biological samples, this soft-release characteristic of desthiobiotin also helps to minimize co-purification of endogenous biotinylated molecules, which remain bound to streptavidin upon elution of the target protein complexes with free biotin. The modified avidin-biotin affinity system also eliminates the need to use harsh elution conditions that might disassociate complexes and/or damage the target protein or cell. Desthiobiotin-based techniques are ideal when using native or recombinant proteins that are not expressed with a fusion tag and when isolating captured proteins under native conditions, such as targeting intact cells or cell surface proteins.
Labeling with Hydrazide Reagents:
Hydrazides and alkoxyamines are two types of carbonyl-reactive groups. Hydrazides (–NH-NH2) react specifically with aldehyde groups in slightly acidic conditions to form hydrazone linkages; these can be further reduced to stable secondary amine bonds using sodium cyanoborohydride (Part No. 44892). The reaction is more efficient in the presence of aniline (Part No. 88944). Alternatively, hydrazides can be conjugated to carboxylic acids using EDC carbodiimide chemistry.
Reactive aldehyde groups can be generated in glycoproteins and other polysaccharide compounds by oxidation of constituent sugar diols using sodium periodiate (Part No. 20504). Sialic acid residues are common components of protein glycosylation and are easily converted to aldehydes with 1mM NaIO4.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Hydrazide-activated desthiobiotin with a tetraethyleneglycol spacer
Product Type EZ-Link Hydrazide-PEG4-Desthiobiotin, No-Weigh Format
Quantity 5 x 1 mg
Sufficient For 5 uses to label a total of 2mg of IgG using typical conditions
Content And Storage 4°C; -20°C as stock solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.