Learn More
Thermo Scientific™ Barnstead™ GenPure™ xCAD Plus Remote Dispensers
Increase flexibility with additional remote dispensers
Brand:  Thermo Scientific™ 50136494
 View more versions of this product
Product Code. 15182883
  • £1497.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in bench and wall-mounted versions
  • Attach up to two additional dispensers per system
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GenPure xCAD Plus, GenPure xCAD Plus UV, GenPure xCAD Plus UF, GenPure xCAD Plus UV/UF, GenPure xCAD Plus UV - TOC, GenPure xCAD Plus UV/UF-TOC
10.2 in.
28.5 in.
21 in.
Water Dispenser, Bench Version
260 mm
725 mm
530 cm