missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Thalidomide-O-amido-PEG3-C2-NH2 TFA
Specifications
Specifications
| Molecular Weight (g/mol) | 620.53 g/mol |
| CAS | 1957236-21-3 |
| Content And Storage | 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture) |
| Shipping Condition | Room temperature |
| Quantity | 1 g |
| Synonym | Cereblon Ligand-Linker Conjugates 3 TFA ∣ E3 ligase Ligand-Linker Conjugates 14 TFA |
| Solubility | DMSO : 50 mg/mL (80.58 mM; Need ultrasonic) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?