missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TGFBR1 (ALK5), GST Tag Recombinant Protein
Human TGFBR1 (ALK5), GST Tag Recombinant Protein
Brand: Invitrogen™ PV5838
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Upon binding TGF-b, TGFBR2 dimerizes with and activates TGFBR1 via phosphorylation leading to downstream phosphorylation of SMADs by TGFBR1.
Specifications
NP_004603.1 | |
62.9 kDa | |
TGFBR1 (ALK5) | |
Human | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Dry Ice | |
Full Length | |
Serine⁄Threonine Kinases |
Kinase Assay | |
1mg | |
STKR Kinase Family | |
GST Tag | |
Ser⁄Thr Kinases: TKL Group | |
Baculovirus | |
Frozen | |
Drug Development |