missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TGFBR1 (ALK5), GST Tag Recombinant Protein
Human TGFBR1 (ALK5), GST Tag Recombinant Protein
£434.00 - £6900.00
Specifications
Accession Number | NP_004603.1 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 62.9 kDa |
Gene Alias | TGFBR1 (ALK5) |
KinaseFamily | STKR Kinase Family |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10479684
|
Invitrogen™
PV5837 |
10μg |
£434.00
Each |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15428713
|
Invitrogen™
PR9075A |
100μg |
£1604.00
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468793
|
Invitrogen™
PV5838 |
1mg |
£6900.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Upon binding TGF-b, TGFBR2 dimerizes with and activates TGFBR1 via phosphorylation leading to downstream phosphorylation of SMADs by TGFBR1.
Specifications
NP_004603.1 | |
62.9 kDa | |
STKR Kinase Family | |
GST Tag | |
Ser⁄Thr Kinases: TKL Group | |
Baculovirus |
Kinase Assay | |
TGFBR1 (ALK5) | |
Human | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Dry Ice |