missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta RIII Antibody (1C5H11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | TGF-beta RIII |
|---|---|
| Clone | 1C5H11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696995
|
Novus Biologicals
NBP2-37418-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18161219
|
Novus Biologicals
NBP2-37418 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGF-beta RIII Monoclonal specifically detects TGF-beta RIII in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
| TGF-beta RIII | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| betaglycan, betaglycan proteoglycan, BGCAN, TGF-beta receptor type 3, TGF-beta receptor type III, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa) | |
| TGFBR3 | |
| IgG1 | |
| Affinity Purified |
| 1C5H11 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Angiogenesis | |
| PBS with 0.05% Sodium Azide | |
| 7049 | |
| Purified recombinant fragment of human TGFBR3 (AA: 147-328 ) expressed in E. Coli. | |
| Primary | |
| 93.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title