missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta RIII Antibody (1C5H11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37418
This item is not returnable.
View return policy
Description
TGF-beta RIII Monoclonal specifically detects TGF-beta RIII in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| TGF-beta RIII | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| betaglycan, betaglycan proteoglycan, BGCAN, TGF-beta receptor type 3, TGF-beta receptor type III, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa) | |
| Mouse | |
| 93.4 kDa | |
| 0.1 mL | |
| Angiogenesis | |
| 7049 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1C5H11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| TGFBR3 | |
| Purified recombinant fragment of human TGFBR3 (AA: 147-328 ) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction