missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta RII Antibody (RM0132-6G11) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£171.00 - £444.00
Specifications
| Antigen | TGF-beta RII |
|---|---|
| Clone | RM0132-6G11 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18223592
|
Novus Biologicals
NBP1-22508-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18260770
|
Novus Biologicals
NBP1-22508 |
0.1 mg |
£444.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGF-beta RII Monoclonal specifically detects TGF-beta RII in Mouse samples. It is validated for Western Blot.Specifications
| TGF-beta RII | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Angiogenesis, Cell Cycle and Replication | |
| Q62312 | |
| 7048 | |
| IgG2 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| RM0132-6G11 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbeta-RII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
| TGFBR2 | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title