missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta RII Antibody (RM0132-6G11) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22508
This item is not returnable.
View return policy
Description
TGF-beta RII Monoclonal specifically detects TGF-beta RII in Mouse samples. It is validated for Western Blot.
Specifications
| TGF-beta RII | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000 | |
| Q62312 | |
| TGFBR2 | |
| Mouse recombinant TGF-beta RII (extracellular domain) | |
| 0.1 mg | |
| Angiogenesis, Cell Cycle and Replication | |
| 7048 | |
| Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
| Western Blot | |
| RM0132-6G11 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbeta-RII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody does not cross-react with mouse TGF-beta RI. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction