missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat TfR (Transferrin R) Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
£241.00 - £448.00
Specifications
| Antigen | TfR (Transferrin R) |
|---|---|
| Clone | OX26 |
| Dilution | Flow Cytometry 0.5 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18742408
![]() |
R&D Systems
FAB9294R-025UG |
25 μg |
£241.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18723338
![]() |
R&D Systems
FAB9294R-100UG |
100 μg |
£448.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Rat samples. It is validated for Flow Cytometry.Specifications
| TfR (Transferrin R) | |
| Flow Cytometry 0.5 ug/10^6 cells | |
| Monoclonal | |
| Mouse | |
| Rat | |
| CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
| PHA activated rat lymphocytes | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| OX26 | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| 7037 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects rat TfR (Transferrin R) in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title