missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat TfR (Transferrin R) Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9294R-025UG
This item is not returnable.
View return policy
Description
TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Rat samples. It is validated for Flow Cytometry.
Specifications
| TfR (Transferrin R) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7037 | |
| Rat | |
| IgG2a |
| Flow Cytometry | |
| OX26 | |
| Flow Cytometry 0.5 ug/10^6 cells | |
| CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
| PHA activated rat lymphocytes | |
| 25 μg | |
| Primary | |
| Detects rat TfR (Transferrin R) in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction