missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TfR (Transferrin R) Antibody (1A1B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00 - £435.00
Specifications
| Antigen | TfR (Transferrin R) |
|---|---|
| Clone | 1A1B2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18160419
|
Novus Biologicals
NBP2-37317 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657005
|
Novus Biologicals
NBP2-37317-0.025ml |
0.025 mL |
£196.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, ELISA.Specifications
| TfR (Transferrin R) | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Monkey, Primate | |
| CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
| TFRC | |
| IgG1 | |
| Affinity Purified |
| 1A1B2 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 7037 | |
| Purified recombinant fragment of human TFRC (AA: 608-727) expressed in E. Coli. | |
| Primary | |
| 85 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title