missing translation for 'onlineSavingsMsg'
Learn More

TfR (Transferrin R) Antibody (1A1B2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-37317-0.025ml

 View more versions of this product

Product Code. 18657005

  • £196.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TfR (Transferrin R)
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Mouse
85 kDa
0.025 mL
Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
7037
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Western Blot, ELISA
1A1B2
Unconjugated
PBS with 0.05% Sodium Azide
TFRC
Purified recombinant fragment of human TFRC (AA: 608-727) expressed in E. Coli.
Affinity Purified
RUO
Primary
Human, Mouse, Rat, Monkey, Primate
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only