missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TFPI Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £474.00
Specifications
| Antigen | TFPI |
|---|---|
| Clone | 374718 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18711899
![]() |
R&D Systems
MAB29741-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18126141
|
R&D Systems
MAB29741 |
500 μg |
£474.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFPI Monoclonal specifically detects TFPI in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| TFPI | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| convertin, EPI, EPITFPI1LACITFI, Extrinsic pathway inhibitor, LACI, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| TFPI | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 374718 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P10646 | |
| 7035 | |
| Mouse myeloma cell line NS0-derived recombinant human TFPI Asp29-Lys282 Accession # P10646 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TFPI in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Coagulation factor II, VII, XI, XIV, rhTFPI-2, recombinant mouse (rm) TFPI, rmTFPI-2, or rhVitronectin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title