missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TFPI Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB29741-SP
This item is not returnable.
View return policy
Description
TFPI Monoclonal specifically detects TFPI in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| TFPI | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| convertin, EPI, EPITFPI1LACITFI, Extrinsic pathway inhibitor, LACI, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7035 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 374718 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P10646 | |
| TFPI | |
| Mouse myeloma cell line NS0-derived recombinant human TFPI Asp29-Lys282 Accession # P10646 | |
| 25 μg | |
| Primary | |
| Detects human TFPI in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Coagulation factor II, VII, XI, XIV, rhTFPI-2, recombinant mouse (rm) TFPI, rmTFPI-2, or rhVitronectin is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction