missing translation for 'onlineSavingsMsg'
Learn More

Ebro Electronic™ TFH 610 / TFH 620 Thermo-Hygrometer series

Thermo-Hygrometer for humidity and temperature measurement.

Brand:  Ebro Electronic™ 1340-5621A

Code : A0

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 10402850

  • £368.00 / Each
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • robust and impact resistant
  • High accuracy
  • °C/°F selectable
  • Approximately 5 years battery life time
  • Automatic deactivation
Specifications

Specifications

Thermohygrometer
0.1% rH
0°C to 60°C
0.1°C
90 g
0 to 100%
±2% rH
±0.3°C
115 x 54 x 22 mm
Factory calibration certificate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers