missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEB Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
£180.00 - £409.00
Specifications
| Antigen | TFEB |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18218070
|
Novus Biologicals
NB100-93447 |
0.1 mg |
£409.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18509433
|
Novus Biologicals
NB100-93447-20ug |
20 μg |
£180.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFEB Polyclonal specifically detects TFEB in Human, Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
| TFEB | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
| TFEB | |
| IgG | |
| Affinity Purified |
| 0.5 mg/mL | |
| Polyclonal | |
| Goat | |
| Human, Rat | |
| P19484 | |
| 7942 | |
| Peptide with sequence C-QKDLQKSRELENH corresponding to internal region according to NP_009093.1. | |
| Primary | |
| This antibody is expected not to cross-react to TFE3 and MITF. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title