missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEB Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-93447
This item is not returnable.
View return policy
Description
TFEB Polyclonal specifically detects TFEB in Human, Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| TFEB | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7942 | |
| Human, Rat | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:1000 | |
| P19484 | |
| TFEB | |
| Peptide with sequence C-QKDLQKSRELENH corresponding to internal region according to NP_009093.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected not to cross-react to TFE3 and MITF. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction