missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Texas Red™-X, Succinimidyl Ester, mixed isomers

Product Code. 11510796
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11510796 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11510796 Supplier Invitrogen™ Supplier No. T6134
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Texas Red™-X, Succinimidyl Ester, mixed isomers

The isomeric mixture of amine-reactive Texas Red-X, succinimidyl ester and its conjugates emit at a longer wavelength than do either tetramethylrhodamine or Lissamine rhodamine B, and are among the most commonly used long-wavelength third labels in fluorescence microscopy and flow cytometry. Protein conjugates prepared with Texas Red-X succinimidyl ester also have a higher fluorescence yield than those with the same labeling ratio prepared with Texas Red sulfonyl chloride.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Texas Red™
Label Type Classic Dyes
Product Line Texas Red

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.