missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphthalic anhydride, 97+%

Product Code. 11414044
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414044 5 g 5g
11349216 10 g 10g
11424044 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414044 Supplier Thermo Scientific Alfa Aesar Supplier No. A11998.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4741-53-1 | C32H20O3 | 452.509 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4741-53-1
Molecular Formula C32H20O3
Molecular Weight (g/mol) 452.509
MDL Number MFCD00044504
InChI Key RSKXGCFFFZIWNC-UHFFFAOYSA-N
Synonym tetraphenylphthalic anhydride, 4,5,6,7-tetraphenylisobenzofuran-1,3-dione, acmc-20ajiw, tetraphenylphthalicanhydride, 3,4,5,6-tetraphenylphthalic anhydride, tetraphenyl-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, 4,5,6,7-tetraphenyl
PubChem CID 78478
IUPAC Name 4,5,6,7-tetraphenyl-2-benzofuran-1,3-dione
SMILES C1=CC=C(C=C1)C2=C(C(=C3C(=C2C4=CC=CC=C4)C(=O)OC3=O)C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 289°C to 297°C
Quantity 5 g
Beilstein 353504
Sensitivity Moisture sensitive
Formula Weight 452.51
Percent Purity 98%
Chemical Name or Material Tetraphenylphthalic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.