CAS RN 4741-53-1
IUPAC Name:
tetraphenyl-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms:
tetraphenyl-2-benzofuran-1,3-dione
Molecular Weight (g/mol):
452.51
Molecular Formula:
C32H20O3
InChi Key:
RSKXGCFFFZIWNC-UHFFFAOYSA-N
SMILES:
O=C1OC(=O)C2=C1C(C1=CC=CC=C1)=C(C1=CC=CC=C1)C(C1=CC=CC=C1)=C2C1=CC=CC=C1
1
–
1
of
1
results
1
Tetraphenylphthalic anhydride, 97+%
CAS: 4741-53-1 Molecular Formula: C32H20O3 Molecular Weight (g/mol): 452.509 MDL Number: MFCD00044504 InChI Key: RSKXGCFFFZIWNC-UHFFFAOYSA-N Synonym: tetraphenylphthalic anhydride,4,5,6,7-tetraphenylisobenzofuran-1,3-dione,acmc-20ajiw,tetraphenylphthalicanhydride,3,4,5,6-tetraphenylphthalic anhydride,tetraphenyl-2-benzofuran-1,3-dione,1,3-isobenzofurandione, 4,5,6,7-tetraphenyl PubChem CID: 78478 IUPAC Name: 4,5,6,7-tetraphenyl-2-benzofuran-1,3-dione SMILES: C1=CC=C(C=C1)C2=C(C(=C3C(=C2C4=CC=CC=C4)C(=O)OC3=O)C5=CC=CC=C5)C6=CC=CC=C6