missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tetramethylrhodamine-5-Isothiocyanate (5-TRITC; G isomer)
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
The amine-reactive tetramethylrhodamine-5-isothiocyanate (5-TRITC; G isomer) can be used to can be used to create bright red-orange fluorescent bioconjugates with excitation/emission maxima ∼555/580 nm.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Tetramethylrhodamine-5-isothiocyanate |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Isothiocyanate |
| Label or Dye | TMR (Tetramethylrhodamine), TRITC Isomers |
| Label Type | Classic Dyes |
| Emission | 580 |
| Excitation | 555 |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction