missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(diethylamino)titanium(IV), 99.99% (metals basis), Thermo Scientific™

Product Code. 11328438
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328438 2 g 2g
11318438 10 g 10g
11338438 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328438 Supplier Thermo Scientific Alfa Aesar Supplier No. 040469.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Nitride precursor - MOCVD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4419-47-0
Molecular Formula C16H40N4Ti
Molecular Weight (g/mol) 336.391
MDL Number MFCD00015648
InChI Key VJDVOZLYDLHLSM-UHFFFAOYSA-N
Synonym tetrakis diethylamino titanium, tetrakis diethylamino titanium iv, ethanamine, n-ethyl-, titanium 4+ salt 4:1, titanium tetrakis diethylamide, tdeat, acmc-20ajjg, ti net2 4, titanium, tetrakis diethylamino, tetrakis diethylamido titanium iv, titanium iv tetrakis diethylamide
PubChem CID 123216
IUPAC Name diethylazanide;titanium(4+)
SMILES CC[N-]CC.CC[N-]CC.CC[N-]CC.CC[N-]CC.[Ti+4]

Specifications

Color Orange to Red
Density 0.95g/mL
Boiling Point 133°C (1.2 mmHg)
Linear Formula Ti[N(C2H5)2]4
Refractive Index 1.5370
Quantity 2 g
UN Number UN3129
Sensitivity Air & Moisture Sensitive
Formula Weight 336.4
Percent Purity 99.99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tetrakis(diethylamino)titanium(IV)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.