accessibility menu, dialog, popup

UserName

TiCH3NH3CCH3NH3CCH3NH3CCH3NH3C

CAS RN 4419-47-0

IUPAC Name: titanium(4+) tetrakis(diethylazanide)
Synonyms: titanium(4+) tetrakis(diethylazanide)
Molecular Weight (g/mol): 336.39
Molecular Formula: C16H40N4Ti
InChi Key: VJDVOZLYDLHLSM-UHFFFAOYSA-N
SMILES: [Ti+4].CC[N-]CC.CC[N-]CC.CC[N-]CC.CC[N-]CC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)