missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(decyl)ammonium bromide, 98%, Thermo Scientific Chemicals
£90.70 - £344.00
Chemical Identifiers
CAS | 14937-42-9 |
---|---|
Molecular Formula | C40H84BrN |
Molecular Weight (g/mol) | 659.01 |
InChI Key | AHNISXOXSNAHBZ-UHFFFAOYSA-M |
Synonym | tetrakis decyl ammonium bromide, tetra decyl ammonium bromide, tetra-decylammonium bromide, tetra-n-decylammonium bromide, acmc-1c6fh, tetrakis-decylazanium bromide, tetrakis decyl azanium bromide, 1-decanaminium, n,n,n-tris decyl-, bromide |
PubChem CID | 3014876 |
IUPAC Name | tetrakis-decylazanium;bromide |
SMILES | CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC.[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11995451
|
Thermo Scientific Acros
435670100 |
10 g | Glass bottle |
£90.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11905461
|
Thermo Scientific Acros
435670500 |
50 g | Glass bottle |
£344.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
14937-42-9 | |
659.01 | |
tetrakis decyl ammonium bromide, tetra decyl ammonium bromide, tetra-decylammonium bromide, tetra-n-decylammonium bromide, acmc-1c6fh, tetrakis-decylazanium bromide, tetrakis decyl azanium bromide, 1-decanaminium, n,n,n-tris decyl-, bromide | |
tetrakis-decylazanium;bromide |
C40H84BrN | |
AHNISXOXSNAHBZ-UHFFFAOYSA-M | |
3014876 | |
CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC.[Br-] |
Specifications
88.0°C | |
White | |
97.5% min. (Argentometry) | |
tetrakis decyl ammonium bromide, tetra decyl ammonium bromide, tetra-decylammonium bromide, tetra-n-decylammonium bromide, acmc-1c6fh, tetrakis-decylazanium bromide, tetrakis decyl azanium bromide, 1-decanaminium, n,n,n-tris decyl-, bromide | |
CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC.[Br-] | |
659.01 | |
659.01 | |
Authentic |
14937-42-9 | |
Fine Powder | |
C40H84BrN | |
AHNISXOXSNAHBZ-UHFFFAOYSA-M | |
tetrakis-decylazanium;bromide | |
3014876 | |
98% | |
Tetrakis(decyl)ammonium bromide |
Safety and Handling
missing translation for 'einecsNumber' : 239-013-3
RUO – Research Use Only