missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(decyl)ammonium bromide, 98%

Product Code. 11995451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11995451 10 g Glass bottle 10g
11905461 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11995451 Supplier Thermo Scientific Acros Supplier No. 435670100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14937-42-9 | C40H84BrN | 659.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14937-42-9
Molecular Formula C40H84BrN
Molecular Weight (g/mol) 659.01
InChI Key AHNISXOXSNAHBZ-UHFFFAOYSA-M
Synonym tetrakis decyl ammonium bromide, tetra decyl ammonium bromide, tetra-decylammonium bromide, tetra-n-decylammonium bromide, acmc-1c6fh, tetrakis-decylazanium bromide, tetrakis decyl azanium bromide, 1-decanaminium, n,n,n-tris decyl-, bromide
PubChem CID 3014876
IUPAC Name tetrakis-decylazanium;bromide
SMILES CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC.[Br-]

Specifications

Melting Point 88.0°C
Color White
Physical Form Fine Powder
Quantity 10 g
Assay Percent Range 97.5% min. (Argentometry)
Formula Weight 659.01
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Tetrakis(decyl)ammonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.