missing translation for 'onlineSavingsMsg'
Learn More

Tetraisopropyl Dichloromethylene Diphosphonate, TRC

Product Code. 30411244
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30411244 50 mg
30416307 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30411244 Supplier Toronto Research Chemicals Supplier No. TRCT29622550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10596-22-2
Molecular Formula C13 H28 Cl2 O6 P2
Molecular Weight (g/mol) 413.21
IUPAC Name 2-[[dichloro-di(propan-2-yloxy)phosphorylmethyl]-propan-2-yloxyphosphoryl]oxypropane
SMILES CC(C)OP(=O)(OC(C)C)C(Cl)(Cl)P(=O)(OC(C)C)OC(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H28Cl2O6P2/c1-9(2)18-22(16,19-10(3)4)13(14,15)23(17,20-11(5)6)21-12(7)8/h9-12H,1-8H3
Formula Weight 412.0738
Chemical Name or Material Tetraisopropyl Dichloromethylene Diphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.