CAS RN 10596-22-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tetraisopropyl Dichloromethylene Diphosphonate, TRC
CAS: 10596-22-2 Molecular Formula: C13 H28 Cl2 O6 P2 Molecular Weight (g/mol): 413.21 IUPAC Name: 2-[[dichloro-di(propan-2-yloxy)phosphorylmethyl]-propan-2-yloxyphosphoryl]oxypropane SMILES: CC(C)OP(=O)(OC(C)C)C(Cl)(Cl)P(=O)(OC(C)C)OC(C)C