missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromo-p-benzoquinone, 95%, Thermo Scientific™

Product Code. 10076322 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10076322 1 g Glass bottle 1g
10737913 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10076322 Supplier Thermo Scientific Acros Supplier No. 386190010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-48-2
Molecular Formula C6Br4O2
Molecular Weight (g/mol) 423.68
MDL Number MFCD00013785
InChI Key LWHDQPLUIFIFFT-UHFFFAOYSA-N
Synonym bromanil, tetrabromo-1,4-benzoquinone, bromanyl, p-bromanil, tetrabromoquinone, tetrabromo-p-benzoquinone, p-bromoanil, tetrabromo-p-quinone, tetrabromobenzoquinone, p-quinone, tetrabromo
PubChem CID 68101
SMILES BrC1=C(Br)C(=O)C(Br)=C(Br)C1=O

Specifications

Melting Point 290°C to 297°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula C6Br4(=O)2
Quantity 1 g
IUPAC Name 2,3,5,6-tetrabromocyclohexa-2,5-diene-1,4-dione
Formula Weight 423.68
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromo-p-benzoquinone
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabromo-p-benzoquinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.