CAS RN 488-48-2
IUPAC Name:
tetrabromocyclohexa-2,5-diene-1,4-dione
Synonyms:
bromanyl
Molecular Weight (g/mol):
423.68
Molecular Formula:
C6Br4O2
InChi Key:
LWHDQPLUIFIFFT-UHFFFAOYSA-N
SMILES:
BrC1=C(Br)C(=O)C(Br)=C(Br)C1=O
1
–
3
of
3
results
1
Bromanil, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bromanil, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tetrabromo-p-benzoquinone, 95%, Thermo Scientific™
CAS: 488-48-2 Molecular Formula: C6Br4O2 Molecular Weight (g/mol): 423.68 MDL Number: MFCD00013785 InChI Key: LWHDQPLUIFIFFT-UHFFFAOYSA-N Synonym: bromanil,tetrabromo-1,4-benzoquinone,bromanyl,p-bromanil,tetrabromoquinone,tetrabromo-p-benzoquinone,p-bromoanil,tetrabromo-p-quinone,tetrabromobenzoquinone,p-quinone, tetrabromo PubChem CID: 68101 SMILES: BrC1=C(Br)C(=O)C(Br)=C(Br)C1=O