missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium hydrogen sulfate, 98%

Product Code. 11414054
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414054 1 g 1g
11424054 5 g 5g
11434054 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414054 Supplier Thermo Scientific Alfa Aesar Supplier No. A12002.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 56211-70-2 | C12H29NO4S | 283.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56211-70-2
Molecular Formula C12H29NO4S
Molecular Weight (g/mol) 283.43
MDL Number MFCD00038764
InChI Key MOXJKKOSZCHGEU-UHFFFAOYSA-M
Synonym tetrapropylammonium bisulfate, tetra-n-propylammonium hydrogen sulfate, tetrapropylammonium hydrogen sulfate, hydrogen sulfate; tetrapropylammonium, tetrapropyl ammonium hydrogen sulphate, acmc-1avly, hydrogen sulfate; tetrapropylazanium, n,n,n-tripropylpropan-1-aminium hydrogen sulfate, 1-propanaminium, n,n,n-tripropyl-, sulfate
PubChem CID 16218669
IUPAC Name hydrogen sulfate;tetrapropylazanium
SMILES OS([O-])(=O)=O.CCC[N+](CCC)(CCC)CCC

Specifications

Melting Point 158°C to 160°C
Linear Formula (CH3CH2CH2)4NHSO4
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 283.43
Percent Purity 98%
Chemical Name or Material Tetra-n-propylammonium hydrogen sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.