accessibility menu, dialog, popup

UserName

CAS RN 56211-70-2

CH3NH3CH3CH3COHSOOO

CAS RN 56211-70-2

IUPAC Name: tetrapropylazanium hydrogen sulfate
Synonyms: tetrapropylammonium hydrogen sulfate
Molecular Weight (g/mol): 283.43
Molecular Formula: C12H29NO4S
InChi Key: MOXJKKOSZCHGEU-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.CCC[N+](CCC)(CCC)CCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)