missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium hydrogen sulfate, 98%

Product Code. 11434054
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434054 25 g 25g
11414054 1 g 1g
11424054 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434054 Supplier Thermo Scientific Alfa Aesar Supplier No. A12002.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56211-70-2 | C12H29NO4S | 283.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56211-70-2
Molecular Formula C12H29NO4S
Molecular Weight (g/mol) 283.43
MDL Number MFCD00038764
InChI Key MOXJKKOSZCHGEU-UHFFFAOYSA-M
Synonym tetrapropylammonium bisulfate, tetra-n-propylammonium hydrogen sulfate, tetrapropylammonium hydrogen sulfate, hydrogen sulfate; tetrapropylammonium, tetrapropyl ammonium hydrogen sulphate, acmc-1avly, hydrogen sulfate; tetrapropylazanium, n,n,n-tripropylpropan-1-aminium hydrogen sulfate, 1-propanaminium, n,n,n-tripropyl-, sulfate
PubChem CID 16218669
IUPAC Name hydrogen sulfate;tetrapropylazanium
SMILES OS([O-])(=O)=O.CCC[N+](CCC)(CCC)CCC

Specifications

Melting Point 158°C to 160°C
Linear Formula (CH3CH2CH2)4NHSO4
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 283.43
Percent Purity 98%
Chemical Name or Material Tetra-n-propylammonium hydrogen sulfate
compliance-icons
Product Identifier
  • Tetra-n-propylammonium hydrogen sulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetra-n-propylammonium hydrogen sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.