missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£177.00 - £366.00
Specifications
| Antigen | TET3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618431
|
Novus Biologicals
NBP2-93429-0.02ml |
0.02 mL |
£177.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608261
|
Novus Biologicals
NBP2-93429-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TET3 Polyclonal antibody specifically detects TET3 in Mouse samples. It is validated for Western BlotSpecifications
| TET3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| PBS (pH 7.3), 50% glycerol | |
| 200424 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1551-1650 of human TET3 (NP_001274420.1). GFQDKLWNPMKGEEGRIPAAGASQLDRAWQSFGLPLGSSEKLFGALKSEEKLWDPFSLEEGPAEEPPSKGAVKEEKGGGGAEEEEEELWSDSEHNFLDEN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title