missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Mouse, Clone: 4E1G2, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio MSM1-250-P1
Additional Details : Weight : 0.01000kg
Specifications
Testosterone | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
Secreted and cytoplasmic | |
100 μg | |
Primary | |
IgG1 κ |
Immunohistochemistry | |
4E1G2 | |
Unconjugated | |
Mouse | |
Protein A/G Purified | |
RUO | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |