missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Mouse, Clone: 4E1G2, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Testosterone |
---|---|
Clone | 4E1G2 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941338
|
EnquireBio
MSM1-250-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931338
|
EnquireBio
MSM1-250-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Testosterone | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
4E1G2 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
IgG1 κ | |
Protein A/G Purified | |
Secreted and cytoplasmic |