missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Test Tube Racks for 5 L Baths

Product Code. 17551906 Shop All Fisherbrand Products
Click to view available options
Holds:
16 x 16 to 19 mm Tubes
24 mm Tubes
30 x 10 to 13 mm Tubes
35 x 1.5 ml Tubes
44 x 0.5 mL Tubes
5 x 30 mm Tubes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17551906

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Test tube racks for use with Fisherbrand™ 5 L Digital Heated Circulating Baths.

  • Conveniently stores test tubes being used with Fisherbrand™ 5 L Digital Heated Circulating Baths
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Test Tubes
Holds 35 x 1.5 ml Tubes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.