missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Test Tube Racks for 5 L Baths
Shop All Fisherbrand Products
Click to view available options
Holds:
16 x 16 to 19 mm Tubes
24 mm Tubes
30 x 10 to 13 mm Tubes
35 x 1.5 ml Tubes
44 x 0.5 mL Tubes
5 x 30 mm Tubes
Unit Size:
Each
Description
- Conveniently stores test tubes being used with Fisherbrand™ 5 L Digital Heated Circulating Baths
Specifications
Specifications
| For Use With (Equipment) | Test Tubes |
| Holds | 24 mm Tubes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction