missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Test Tube Rack

OHAUS™ Test Tube Rack designed to securely hold tubes in place.

Brand:  OHAUS™ 30400118

Additional Details : Weight : 1.40000kg

 View more versions of this product

Product Code. 15702539

  • £160.00 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Constructed of stainless steel
  • For use with tubes of various sizes
Specifications

Specifications

Test Tube Rack
Tubes (22 to 25 mm)
24
12.7 x 1.8 x 10.2 cm
5.00 in.
7.00 in.
4.00 in.
12.70 cm
Test Tube Rack 22-25 mm Diameter
PVC-coated Steel
22 to 25 mm
12.70 cm
17.80 cm
10.200 cm
5.00 in.
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers