Learn More
Terumo™ Stainless Steel Needles
Use Teramo™ Stainless Steel Needles for research. Each single-use needle has a color-coded hub and a beveled tip. Their use is for the ZR-205 and SZR-260 series.
Brand: Henry Schein Medical TUAN2332R1
Code : 02
Additional Details : Weight : 0.02000kg
Description
- For SZR-205 and SZR-260 series
- Stainless steel with color-coded polypropylene luer fitting
- Beveled tip
- Sterilized by ethylene oxide
Specifications
Stainless Steel | |
23 G | |
For research use only, single use with colour coded hubs | |
Bevel | |
Sterile |
Blue | |
0.5 mm | |
Needle | |
SZR-205 and SZR-260 Series |