missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 10565110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10565110 1 g Glass Bottle 1g
10210130 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10565110 Supplier Thermo Scientific Acros Supplier No. 438020010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 365996-05-0
Molecular Formula C11H17N3O2S
Molecular Weight (g/mol) 255.34
InChI Key BMLHPGOMLGKYIJ-UHFFFAOYSA-N
Synonym tert-butyl 2-amino-6,7-dihydrothiazolo 5,4-c pyridine-5 4h-carboxylate, tert-butyl 2-amino-6,7-dihydro 1,3 thiazolo 5,4-c pyridine-5 4h-carboxylate, 5-boc-2-amino-4,5,6,7-tetrahydrothiazolo 5,4-c pyridine, 2-amino-6,7-dihydro-4h-thiazolo 5,4-c pyridine-5-carboxylic acid tert-butyl ester, tert-butyl 2-amino-4h,5h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, 2-amino-5-boc-6,7-dihydro-4h-thiazolo-5,4,c pyridine, tert-butyl 2-amino-4h,6h,7h-1,3 thiazolo 5,4-c pyridine-5-carboxylate, tert-butyl 2-amino-6,7-dihydro-4h-thiazolo 5,4-c pyridine-5-carboxylate, pubchem17893
PubChem CID 11357283
IUPAC Name tert-butyl 2-amino-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2=C(C1)SC(=N2)N

Specifications

Melting Point 223.5°C to 224.5°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 255.34
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate
compliance-icons
Product Identifier
  • tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.