accessibility menu, dialog, popup

UserName

CH3H3CCH3OONSNH2N

CAS RN 365996-05-0

IUPAC Name: tert-butyl 2-amino-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
Synonyms: tert-butyl 2-amino-4H,6H,7H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate
Molecular Weight (g/mol): 255.34
Molecular Formula: C11H17N3O2S
InChi Key: BMLHPGOMLGKYIJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC2=C(C1)SC(N)=N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)