Learn More
Thermo Scientific™ Tergitol™ 7-Lactose-TTC Agar
Product Code. 13235549
Click to view available options
Packaging Type:
Boxes with 2 x 10 plates wrapped in film
Flow wrapped - Single layer
Unit Size:
Pack of 10
Pack of 100
Product Code. 13235549
Brand: Thermo Scientific™ PO5424J
Enumerate and differentiate coliforms from water and food samples with this ready-to-use medium.
Enumerate and differentiate coliforms from water and food samples with ready-to-use Thermo Scientific™ Oxoid™ Tergitol™ 7-Lactose-TTC Agar. Tergitol 7 inhibits Gram-positive organisms allowing superior recovery of coliforms. The addition of tri-phenyltetrazolium chloride (TTC) allows recognition and identification of Escherichia coli and Enterobacter aerogenes. Fermentation of lactose is seen by a change in colour of the pH indicator bromothymol blue. TTC is rapidly reduced to red formazan by most coliform organisms except E. coli and Enterobacter aerogenes allowing easy differentiation.
Use Tergitol 7-Lactose-TTC Agar for the detection and enumeration of coliforms in food and water samples.
  • Easy-to-read: easily differentiate Escherichia coli (yellow colonies) and Enterobacter species (greenish/yellow colonies) from coliforms (red colonies).
  • Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Tergitol™ is a trademark of the Dow Chemical Company (Dow)
TRUSTED_SUSTAINABILITY
Specifications
Format 55 mm Plate
Product Type Agar
Fill Weight 3.2 ± 0.2
Packaging Type Flow wrapped - Single layer